Key Documents
CLS6970SR
Corning® magnetic stir bar retrievers
ergonomic stir bar retriever, 1/cs
Synonym(s):
stir bar retrievers
About This Item
UNSPSC Code:
41100000
NACRES:
NB.73
Pricing and availability is not currently available.
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 6970SR
compatibility
all stirring models
General description
The Corning stir bar retriever contains a strong magnet to remove a stir bar from all glass and plastic containers used in the laboratory. The container must be taken off the magnetic stirrer before using the retriever as the high-powered magnet in the stirrer cancels out the one in the retriever. The handle is designed for ergonomic and easy retrieval. The retriever is available one per case.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)