CLS6406250FO
PYREX® replacement Squibb separatory funnel w/o valve and stopper
pack of 4 ea
Synonym(s):
equipment replacement accessory
All Photos(1)
About This Item
packaging
pack of 4 ea
manufacturer/tradename
Corning 6406-250FO
capacity
250 mL
volume
250 mL
General description
These economy Squibb separatory funnel replacements do not have a Polytetrafluorethylene (PTFE) valve assembly or polyethylene stopper.
Legal Information
PYREX is a registered trademark of Corning, Inc.