Skip to Content
Merck
All Photos(1)

Key Documents

CLS3826

Corning® 384 well microplate, low volume

Tissue Culture (TC)-treated surface, white polystyrene, flat bottom, sterile, lid, pack of 10

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
flat bottom
white bottom
white polystyrene
size:
384 well
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid
skirt
Pricing and availability is not currently available.

material

flat bottom
white bottom
white polystyrene

sterility

sterile

feature

lid
skirt

packaging

pack of 10
case of 50

manufacturer/tradename

Corning 3826

size

384 well

surface area

0.03 cm2 , cell growth area

well volume

50 μL

well working volume

5-40 μL

wells

384 wells

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Round well design with a solid white flat bottom.
  • Surface treated for enhanced cell attachment.
  • Ideal for miniaturized 384 well fluorescent cell-based assays

Note: Suitable for 384 well fluorescent cell-based assays

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service