CLS2980223
Corning® cover glasses
rectangular, No. 11/2, W × L 22 mm × 30 mm
Synonym(s):
coverslips, glass covers, slide covers
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.16
Recommended Products
material
borosilicate glass
rectangular
packaging
pack of 100 ea
case of 1000 ea
manufacturer/tradename
Corning 2980-223
W × L
22 mm × 30 mm
binding type
non-treated surface
Related Categories
Features and Benefits
- Corning® cover glasses are made from optically clear borosilicate glass.
- Chemically resistant cover glasses are not affected by weathering or surface attachment.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)