Skip to Content
Merck
All Photos(1)

Key Documents

B16-F1 Cell Line from mouse

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

mouse skin

packaging

tube of 5 μg 92101203-DNA-5UG
pkg of vial of cells 92101203-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Melanin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse melanoma, producing melanin

Cell Line Description

Mouse melanoma which produces melanin.

Application

B16-F1 cell line from mouse has been used to study the cytotoxic and antioxidant properties of supercritical CO2 extract (SCE) of old man′s beard (Usnea barbata). It has also been used to study the antiproliferative activity of essential oils from Achillea ligustica.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service