Skip to Content
Merck
All Photos(1)

Key Documents

RAMOS (RA.1)

91030710, human lymph, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RAMOS (RA.1), 91030710

biological source

human lymph

growth mode

Suspension

karyotype

Modal no. 45 hypodiploid

morphology

Lymphoblast

products

Not specified

receptors

IL-4, low affinity IgE (CD23)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian Burkitt′s lymphoma

Cell Line Description

This cell line is EBV-genome-negative but expresses EBV receptors and can be converted into a permanent EBV-positive line.

Application

Tumourigenicity and virus studies: EBV

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service