Skip to Content
Merck
All Photos(1)

Documents

BCL1 Clone CW13.20-3B3

90061904, mouse blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

BCL1 Clone CW13.20-3B3, 90061904

biological source

mouse blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c B cell leukaemia

Cell Line Description

Derived from BCL1 by limiting dilution and does not spontaneously secrete IgM when cultured in pre-tested batches of FBS. The line is useful for studying soluble factors involved in B-cell differentiation.

Application

B-cell differentiation studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 1% Sodium Pyruvate (NaP) + 1% Non Essential Amino Acids (NEAA) + 0.05mM 2-Mercaptoethanol (2ME) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells attach lightly to surface which should be tapped sharply to dislodge the cells. Do not allow cells to become confluent, subculture at 75% to 90% confluency; free-floating aggregates acc

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service