Skip to Content
Merck
All Photos(1)

Key Documents

GH3

87012603, rat brain, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

GH3, 87012603

biological source

rat brain

growth mode

Adherent

karyotype

Hyperdiploid, modal no. 60

morphology

Epithelial

products

Growth hormone, prolactin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Wistar-Furth pituitary tumour

Cell Line Description

Derived from a 7 month female Wistar-Furth rat. GH3 cells produce growth hormone faster than the GH1 cell line and also produce prolactin. Hydrocortisone stimulates growth hormone production and inhibits prolactin production. Studies have shown that cells are capable of adaptation to suspension culture using MEM.

Application

Hormone studies

Culture Medium

Ham′s F10 + 2mM Glutamine + 15% Horse Serum (HS) + 2.5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Upon thawing, the time taken before the first subculture can be performed may be extended (10-15 days). To aid attatchment, flask

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service