Skip to Content
Merck
All Photos(1)

Key Documents

BL-3

86062401, bovine blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

BL-3, 86062401, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

bovine blood

packaging

tube of 5 μg 86062401-DNA-5UG
pkg of vial of cells 86062401-1VL

growth mode

Suspension

karyotype

2n = 60, aneuploid, modal no. 65

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Bovine leukaemic lymphoblast

Cell Line Description

BL-3 was established from a neoplastic lymph node of a 3 month old male calf. BVDV positive.

Application

Virus studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 5-9x100,000 cells/ml; 5% CO2; 37°C. Cells should not be over diluted.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service