C-26501
Follicle Dermal Papilla Cell Growth Medium
Ready-to-use kit including Basal Medium and SupplementMix, 500 ml
Synonym(s):
Cell Growth Medium
About This Item
Recommended Products
form
liquid
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
General description
Application
Components
Quality
Preparation Note
Storage and Stability
Subculture Routine
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Primary human follicle dermal papilla cells and optimized culture medium. Protocols for handling, cell passaging, media, and product use.
Primary human follicle dermal papilla cells and optimized culture medium. Protocols for handling, cell passaging, media, and product use.
Primary human follicle dermal papilla cells and optimized culture medium. Protocols for handling, cell passaging, media, and product use.
Primary human follicle dermal papilla cells and optimized culture medium. Protocols for handling, cell passaging, media, and product use.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service