C0495
CDK2/Cyclin A2, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
CCN1, CCNA, p33 (CDK2)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
778-1052 nmol/min·mg
mol wt
CDK2 subunit ~58 kDa
cyclin A2 ~78 kDa
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... CCNA2(890) , CDK2(1017)
Biochem/physiol Actions
CDK2 is a member of the Cyclin-Dependent Kinase family that is ubiquitously expressed. CDK2 is a catalytic subunit of the cyclin-dependent protein kinase complex, whose activity is restricted to the G1-S phase, and essential for cell cycle G1/S phase transition. CDK2 associates with and is regulated by the regulatory subunits of the complex including Cyclin A or E, CDK inhibitor p21Cip1 (CDKN1A) and p27Kip1 (CDKN1B).CDK2 phosphorylates multiple cellular substrates including SMAD3 and FOXO1. Phosphorylation of FOXO1 leads to its inhibition.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protein science : a publication of the Protein Society, 30(7), 1326-1336 (2021-01-17)
In Parkinson's disease with dementia, up to 50% of patients develop a high number of tau-containing neurofibrillary tangles. Tau-based pathologies may thus act synergistically with the α-synuclein pathology to confer a worse prognosis. A better understanding of the relationship between
Science (New York, N.Y.), 314(5797), 294-297 (2006-10-14)
The function of cyclin-dependent kinase 2 (CDK2) is often abolished after DNA damage. The inhibition of CDK2 plays a central role in DNA damage-induced cell cycle arrest and DNA repair. However, whether CDK2 also influences the survival of cells under
Molecular cell, 1(4), 553-563 (1998-07-14)
Apoptosis of human endothelial cells after growth factor deprivation is associated with rapid and dramatic up-regulation of cyclin A-associated cyclin-dependent kinase 2(cdk2) activity. In apoptotic cells, the C termini of the cdk inhibitors p21Cip1/Waf1 and p27Kip1 are truncated by specific
Life science alliance, 4(7) (2021-06-17)
The early-stage pathologies of frontotemporal lobal degeneration (FTLD) remain largely unknown. In VCPT262A-KI mice carrying VCP gene mutation linked to FTLD, insufficient DNA damage repair in neural stem/progenitor cells (NSCs) activated DNA-PK and CDK1 that disabled MCM3 essential for the
Current biology : CB, 31(4), 794-808 (2020-12-29)
The phosphorylation of mitotic proteins is bistable, which contributes to the decisiveness of the transitions into and out of M phase. The bistability in substrate phosphorylation has been attributed to bistability in the activation of the cyclin-dependent kinase Cdk1. However
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service