AXYPCR02LC
MAXYMum Recovery® PCR tubes
with flat caps, thin wall, size 0.2 mL, pkg of 10x1000tubes/cs
Synonym(s):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
About This Item
Recommended Products
material
cap (snap cap)
conical bottom tube
polypropylene
polypropylene cap
manufacturer/tradename
Axygen Inc PCR-02-L-C
Corning
packaging
pkg of 10x1000tubes/cs
size
0.2 mL
color
clear
fitting
cap (push fit)
suitability
suitable for PCR
binding type
(ultra-low retention)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service