AXYMTX1250C
Matrix pipette tips
capacity 1250 μL, pkg of 10x500tips/cs
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
1250 μL
packaging:
pkg of 10x500tips/cs
manufacturer/tradename:
Axygen Inc MTX-1250-C
Corning
Corning
Recommended Products
manufacturer/tradename
Axygen Inc MTX-1250-C
Corning
packaging
pkg of 10x500tips/cs
capacity
1250 μL
volume
1250 μL
Looking for similar products? Visit Product Comparison Guide
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service