Skip to Content
Merck
All Photos(2)

Key Documents

AV44768

Sigma-Aldrich

Anti-PLD3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HU-K4, Anti-Phospholipase D family, member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

mouse, rabbit, bovine, horse, guinea pig, human, rat, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLD3(23646)

Immunogen

Synthetic peptide directed towards the N terminal region of human PLD3

Application

Anti-PLD3 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/ml.

Biochem/physiol Actions

Phospholipase D (PLD) family, member 3 belongs to the PLD family of enzymes that hydrolyze the membrane phospholipids. PLD3 is associated with the endoplasmic reticulum and is expressed in a variety of tissues and cells. The expression of PLD3 increases drastically in neurons and muscle cells during differentiation. PLD3 is involved in the processing of amyloid-beta precursor protein and myogenesis during the formation of the myotube.

Sequence

Synthetic peptide located within the following region: WVLLVLILAVVGFGALMTQLFLWEYGDLHLFGPNQRPAPCYDPCEAVLVE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary Osisami et al.
PloS one, 7(3), e33341-e33341 (2012-03-20)
Phospholipase D3 (PLD3) is a non-classical, poorly characterized member of the PLD superfamily of signaling enzymes. PLD3 is a type II glycoprotein associated with the endoplasmic reticulum, is expressed in a wide range of tissues and cells, and undergoes dramatic
Carlos Cruchaga et al.
Nature, 505(7484), 550-554 (2013-12-18)
Genome-wide association studies (GWAS) have identified several risk variants for late-onset Alzheimer's disease (LOAD). These common variants have replicable but small effects on LOAD risk and generally do not have obvious functional effects. Low-frequency coding variants, not detected by GWAS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service