Skip to Content
Merck
All Photos(1)

Key Documents

A6220

Sigma-Aldrich

N-Acetyl-Ile-Glu-Pro-Asp-7-amido-4-methylcoumarin

≥97% (HPLC), powder

Synonym(s):

Ac-IEPD-AMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H41N5O11
Molecular Weight:
671.69
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

powder

solubility

DMSO/DMF: 1 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(C)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CC(O)=O)C(=O)Nc2ccc3C(C)=CC(=O)Oc3c2

InChI

1S/C32H41N5O11/c1-5-16(2)28(33-18(4)38)31(46)35-21(10-11-25(39)40)32(47)37-12-6-7-23(37)30(45)36-22(15-26(41)42)29(44)34-19-8-9-20-17(3)13-27(43)48-24(20)14-19/h8-9,13-14,16,21-23,28H,5-7,10-12,15H2,1-4H3,(H,33,38)(H,34,44)(H,35,46)(H,36,45)(H,39,40)(H,41,42)/t16-,21-,22-,23-,28-/m0/s1

InChI key

XDMHHBKPMUGLEA-MNTOYCOOSA-N

Amino Acid Sequence

NAc-Ile-Glu-Pro-Asp-AMC

Substrates

Fluorogenic substrate for caspase 8 and granzyme B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Neumann et al.
PloS one, 7(8), e42526-e42526 (2012-08-24)
TNF-related apoptosis-inducing ligand (TRAIL) is a member of the tumor necrosis factor (TNF) ligand family that exerts its apoptotic activity in human cells by binding to two transmembrane receptors, TRAILR1 and TRAILR2. In cells co-expressing both receptors the particular contribution
J L Harris et al.
The Journal of biological chemistry, 273(42), 27364-27373 (1998-10-09)
Granzyme B is a protease involved in the induction of rapid target cell death by cytotoxic lymphocytes. Definition of the substrate specificity of granzyme B allows for the identification of in vivo substrates in this process. By using the combinatorial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service