Skip to Content
Merck
All Photos(1)

Key Documents

A0876

Sigma-Aldrich

N-Acetyl-D-leucine

≥99% (TLC), suitable for ligand binding assays and cell cutlure

Synonym(s):

N-acetyl-D-Leucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO3
CAS Number:
Molecular Weight:
173.21
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

N-Acetyl-D-leucine,

Assay

≥99% (TLC)

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable
ligand binding assay: suitable

color

white

storage temp.

−20°C

SMILES string

CC(C)C[C@@H](NC(C)=O)C(O)=O

InChI

1S/C8H15NO3/c1-5(2)4-7(8(11)12)9-6(3)10/h5,7H,4H2,1-3H3,(H,9,10)(H,11,12)/t7-/m1/s1

InChI key

WXNXCEHXYPACJF-SSDOTTSWSA-N

Application

N-Acetyl-D-leucine may be used with other D-aminoacylated amino acids as a substrate for the identification, differentiation and characterization of D-aminoacylase(s)/amidohydrolase(s).

Biochem/physiol Actions

N-Acetyl-D-leucine is a substrate for D-aminoacylase from Alcaligenes xylosoxydans subsp. xylosoxydans A-6. N-Acetyl-D-leucine is used to help differentiate members of the amidohydrolase enzyme superfamily. It is a preferred substrate of Gox1177 from Gluconobacter oxidans.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Moriguchi et al.
Bioscience, biotechnology, and biochemistry, 57(7), 1149-1152 (1993-07-01)
The best inducers for D-aminoacylase from Alcaligenes xylosoxydans subsp. xylosoxydans A-6 (Alcaligenes A-6) were a poor substrate, N-acetyl-gamma-methyl-D-leucine, and an inhibitor, N-acetyl-D-alloisoleucine. The enzyme has been homogeneously purified. The molecular weight of the native enzyme was estimated to be 58,000
Y B Yang et al.
Bioscience, biotechnology, and biochemistry, 56(9), 1392-1395 (1992-09-01)
The D-aminoacylase produced by Alcaligenes denitrificans DA181 was a new type of aminoacylase which had both high stereospecificity and specific activity. The molecular weight and isoelectric point of this enzyme were 58,000 and 4.4, respectively. The apparent Km and kcat
Jennifer A Cummings et al.
Biochemistry, 48(27), 6469-6481 (2009-06-13)
The catalytic activities of three members of the amidohydrolase superfamily were discovered using amino acid substrate libraries. Bb3285 from Bordetella bronchiseptica, Gox1177 from Gluconobacter oxidans, and Sco4986 from Streptomyces coelicolor are currently annotated as d-aminoacylases or N-acetyl-d-glutamate deacetylases. These three

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service