Skip to Content
Merck
All Photos(1)

Key Documents

902H-0

Sigma-Aldrich

Cytokeratin 5 & 6 (D5/16B4) + TTF-1 (8G7G3/1) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

D5&16B4 + 8G7G3/1, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (902H-07)
bottle of 7.0 mL predilute (902H-08)

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... KRT5(3852)

General description

Anti-Cytokeratin 5 & 6 labels greater than 90% of Eptihelioid Mesotheliomas. Anti-Cytokeratin 5 & 6 stains the cytoplasm of such cells. Anti-TTF-1 stains the nuclei in the case of lung adenocarcinomas and is negative in nearly all mesotheliomas. When the differential diagnosis seeks to distinguish between mesothelioma and adenocarcinoma of the lung, the nuclear vs. Cytoplasmic staining pattern of this cocktail can be of significant value in making the diagnosis.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pablo A Bejarano et al.
Archives of pathology & laboratory medicine, 127(2), 193-195 (2003-02-04)
The immunohistochemical identification of thyroid transcription factor-1 (TTF-1) is regarded as the presence of a nuclear pattern of staining and is used to identify tumors of thyroid or pulmonary origin. Although there have been reports of cytoplasmic expression of TTF-1
Dongfeng Tan et al.
Human pathology, 34(6), 597-604 (2003-06-27)
Thyroid transcription factor 1 (TTF-1), a homeodomain-containing transcription factor, plays a pivotal role in lung development, cell growth, and differentiation processes. The current literature reports considerable variation in frequency of TTF-1 protein expression in human non-small cell lung cancer (NSCLC).
M Srodon et al.
Human pathology, 33(6), 642-645 (2002-08-02)
Metastatic carcinoma of unknown primary origin is a perplexing but common problem, accounting for up to 10% to 15% of all solid tumors at presentation. Many of these metastases presumably arise from primary lung carcinomas, but the morphologic features and
K Y Jang et al.
Analytical and quantitative cytology and histology, 23(6), 400-404 (2002-01-05)
To estimate the utility of thyroid transcription factor-1 (TTF-1) and the combined cytokeratin 7 (CK7) and 20 (CK20) immunoprofile as a marker for identifying the primary site of metastatic adenocarcinoma in effusions of the serous cavity. Formalin-fixed, paraffin-embedded cell block
P M Cury et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 13(2), 107-112 (2000-03-04)
Until recently, the standard approach of most laboratories in distinguishing epithelioid pleural mesothelioma from metastatic adenocarcinoma has been a negative result from a panel of adenocarcinoma-associated antibodies. However, several "mesothelium-associated" antibodies have been proposed as useful in this situation, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service