Skip to Content
Merck
All Photos(1)

Documents

346R-1

Sigma-Aldrich

Villin (EP163) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP163, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (346R-17)
bottle of 7.0 mL predilute (346R-18)
vial of 0.1 mL concentrate (346R-14)
vial of 0.5 mL concentrate (346R-15)
vial of 1.0 mL concentrate (346R-16)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... VIL1(7429)

General description

Villin is a 95 kD glycoprotein of microvilli associated with rootlet formation in gastrointestinal mucosal epithelium. Anti-villin labels the brush border area in the gastrointestinal mucosal epithelium.1 This antibody has been useful in differentiating gastrointestinal adenocarcinoma, neuroendocrine carcinomas, and ovarian adenocarcinomas from adenocarcinomas of other organs.2-5 This antibody also labels Merkel cells of the skin.6

Quality


IVD

IVD

IVD

RUO

Linkage

Villin Positive Control Slides, Product No. 346S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Toyoshima et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 46(11), 1329-1334 (1998-10-17)
Merkel cells represent a population of epithelial cells in the skin and oral mucosa. Although Merkel cells are reliably distinguishable from other epithelial cells at the ultrastructural level, these cells are usually not discernible by standard light microscopy and need
P J Zhang et al.
Archives of pathology & laboratory medicine, 123(9), 812-816 (1999-08-24)
Villin, a 95-kd cytoskeletal protein associated with axial microfilament bundles of brush border microvilli, is mostly restricted to intestinal glandular tumors. Villin immunoexpression was recently observed in a small number of carcinoids of the intestinal tract and lung, but its
N S Goldstein et al.
American journal of clinical pathology, 116(3), 319-325 (2001-09-14)
We studied 14 mucinous and 26 nonmucinous bronchioloalveolar adenocarcinomas (BACs) with thyroid transcription factor (TTF), cytokeratin (CK) 7, CK20, and villin to characterize their staining patterns with these antibodies and identify staining differences between the neoplasms. We also stained 11
J Tan et al.
Human pathology, 29(4), 390-396 (1998-05-01)
Villin (V) is a glycoprotein of microvilli associated with rootlet formation. Most colonic adenocarcinomas have a V positive (+), cytokeratin (CK) 20 (+), CK7-negative (-) immunophenotype; most lung adenocarcinomas have a CK20(-), CK7(+) immunophenotype. The reports of villin immunoreactivity in
Pheroze Tamboli et al.
Archives of pathology & laboratory medicine, 126(9), 1057-1063 (2002-09-03)
To determine whether a limited immunohistochemical panel can help differentiate metastatic colonic adenocarcinoma from primary enteric-type adenocarcinoma of the urinary tract and urothelial (transitional cell) carcinoma with glandular differentiation, which appear morphologically similar but most often necessitate different treatment protocols.

Articles

Colorectal cancer is the third most common cancer in both men and women. An estimated 136,000 cases of colorectal cancer are expected to occur in 2016.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service