Skip to Content
Merck
All Photos(1)

Key Documents

211A-1

Sigma-Aldrich

TSH Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (211A-14)
vial of 0.5 mL concentrate (211A-15)
bottle of 1.0 mL predilute (211A-17)
vial of 1.0 mL concentrate (211A-16)
bottle of 7.0 mL predilute (211A-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

control

pituitary

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Anti-TSH is a useful marker in classification of pituitary tumors and the study of pituitary disease. It reacts with TSH-producing cells (thyrotrophs).

Quality


IVD

IVD

IVD

RUO

Linkage

TSH Positive Control Slides, Product No. 211S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Gessl et al.
The Journal of clinical endocrinology and metabolism, 79(4), 1128-1134 (1994-10-01)
The McCune-Albright syndrome (MAS) comprises a triad of physical signs: localized bone lesions termed polyostotic fibrous dysplasia, café-au-lait pigmentation of the skin, and autonomous hyperfunction of multiple endocrine systems, including overproduction of GH and T4. A somatic activating point mutation
E Batanero et al.
Brain, behavior, and immunity, 6(3), 249-264 (1992-09-01)
We evaluated the presence of anterior pituitary hormones; follicle-stimulating hormone (FSH) and its beta-subunit (beta-FSH), luteinizing hormone (LH) and its beta-subunit (beta-LH), beta-subunit of thyroid-stimulating hormone (beta-TSH), adrenocorticotropic hormone (ACTH), growth hormone (GH), and prolactin (PRL); the placental hormone human
N Sanno et al.
The Journal of clinical endocrinology and metabolism, 80(8), 2518-2522 (1995-08-01)
TSH-secreting pituitary adenomas are rare. The transcriptional expression (messenger ribonucleic acids: mRNAs) of TSH beta, GH, and PRL in five patients with TSH-secreting pituitary adenoma was studied by the in situ hybridization (ISH) method in order to elucidate their multiple
N Kuzuya et al.
The Journal of clinical endocrinology and metabolism, 71(5), 1103-1111 (1990-11-01)
Endocrine and immunohistochemical studies were performed in two cases of TSH-secreting pituitary adenomas. The patients had elevated serum TSH and alpha-subunit concentrations despite high serum thyroid hormone levels. In addition, one patient (no. 1) had elevated serum GH levels with
J N Clore et al.
The American journal of the medical sciences, 295(1), 3-5 (1988-01-01)
A 36-year-old woman with hyperthyroidism, elevated blood thyroid-stimulating hormone (TSH) and alpha-subunit levels, amenorrhea, hyperprolactinemia and no evidence of acromegaly, was found to have a pituitary adenoma containing TSH, alpha-subunit and growth hormone by immunohistochemistry. Preoperative testing revealed elevated TSH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service