Skip to Content
Merck
All Photos(1)

Key Documents

71911

Sigma-Aldrich

Sodium phosphate tribasic dodecahydrate

puriss. p.a., ACS reagent, ≥98.0% (T)

Synonym(s):

TSP, Trisodium phosphate (tert) dodecahydrate, Trisodium phosphate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3PO4 · 12H2O
CAS Number:
Molecular Weight:
380.12
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥98.0% (T)
form:
crystals or chunks

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥98.0% (T)

form

crystals or chunks

impurities

≤0.001% total nitrogen (N)
≤2.5% free alkali (as NaOH)

pH

13 (25 °C, 190.1 g/L)

density

1.62 g/mL at 25 °C

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O

InChI

1S/3Na.H3O4P.12H2O/c;;;1-5(2,3)4;;;;;;;;;;;;/h;;;(H3,1,2,3,4);12*1H2/q3*+1;;;;;;;;;;;;;/p-3

InChI key

ASTWEMOBIXQPPV-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Sodium phosphate tribasic dodecahydrate is a hydrated trisodium salt of phosphoric acid. It is widely used in various water treatment methods.

Application

Sodium phosphate tribasic dodecahydrate (SP tribasic dodecahydrate) may be used to compose buffer solutions (pH in the range of 2 to 12) for use in a study. It may be used to prepare 500mM SP (sodium phosphate) buffer solutions for use in cloud point (CP) experiments.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cloud point phenomenon in amphiphilic drug solutions.
Kim EJ and Shah DO.
Langmuir, 18(26), 10105-10108 (2002)
Inkyu Park et al.
Biosensors & bioelectronics, 22(9-10), 2065-2070 (2006-10-24)
A microneedle sensor platform with integrated silicon nanowire tip was developed for intracellular biochemical detection. Because of the virtue of miniaturized size and high sensitivity, this sensor has a great potential for studying individual cell or localized bioenvironment by revealing
Milne GWA.
Gardner's Commercially Important Chemicals: Synonyms, Trade Names, and Properties, 573-573 (2005)
Jie Mu et al.
Seizure, 23(7), 553-559 (2014-05-24)
To evaluate the value of magnetoencephalography (MEG) source localization in localization of epileptic activities and predicting surgical outcome in frontal lobe epilepsies (FLE). Forty-six patients with presurgical MEG evaluation and intractable FLE surgery (28 male patients) were analyzed retrospectively with
Yuanyuan Li et al.
Chemico-biological interactions, 220, 193-199 (2014-07-12)
Formosanin C (FC) is a pure compound isolated from Rhizoma Paridis. In the past years, antitumor effects of FC have been observed in several cultural cells and animal systems. However, there was no research particular on liver cancer. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service