Skip to Content
Merck
All Photos(1)

Key Documents

27227

Sigma-Aldrich

Ethyl acetate

puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NB.11
Assay:
≥99.5% (GC)
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3 (20 °C, vs air)

Quality Level

vapor pressure

73 mmHg ( 20 °C)

grade

puriss.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

801 °F

quality

meets analytical specification of Ph. Eur., BP, NF

expl. lim.

2.2-11.5 %, 38 °F

impurities

residual solvents, complies
≤0.003% non-volatile matter
≤0.005% free acid (as CH3COOH)
≤0.01% methyl. comp.(as acetic acid methylester)
≤0.1% ethanol (GC)
≤0.1% water (Karl Fischer)
≤0.2% related subst. (GC)

refractive index

n20/D 1.371-1.373
n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

density

0.898-0.902 g/mL at 20 °C
0.894-0.898 g/mL at 25 °C
0.902 g/mL at 25 °C (lit.)

suitability

complies for appearance of solution
complies for reaction against H2SO4
corresponds for chromatography
passes test for identity (IR)

format

neat

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • [Determination of 12 prohibited veterinary drug residues in pig urine by ultra high performance liquid chromatography-tandem mass spectrometry]: Ethyl acetate is utilized for sample preparation in this advanced method to detect drug residues in animal samples, emphasizing its role in veterinary pharmaceutical compliance (Wan et al., 2024).

  • Therapeutic effect of Ginkgetin on smoke-induced airway inflammation by down-regulating the c/EBPβ signaling pathway and CCL2 expression: This study explores the anti-inflammatory properties of Ginkgetin, where ethyl acetate is used in the extraction process, highlighting its utility in biomedical research (Tao et al., 2024).

  • Effect of methanol extract of Plectranthus esculentus N.E.Br tuber and its fractions on indices of benign prostatic hyperplasia in Wistar rats: Ethyl acetate is employed in the extraction of bioactive compounds for pharmacological studies, showing its critical role in therapeutic applications (Kanu et al., 2024).

  • Phytochemical evaluation of Ziziphus mucronata and Xysmalobium undulutum towards the discovery and development of anti-malarial drugs: Demonstrates the use of ethyl acetate in phytochemical screenings, contributing to the development of new anti-malarial medications (Buthelezi et al., 2024).

  • The First Records of the In Silico Antiviral and Antibacterial Actions of Molecules Detected in Extracts of Algerian Fir (Abies numidica De Lannoy) Using LC-MS/MS Analysis: This research utilizes ethyl acetate for extracting compounds from plants for antiviral and antibacterial testing, underscoring its importance in the study of natural products (Benouchenne et al., 2024).

Other Notes

The article number 27227-4X2.5L-M will be discontinued. Please order the single bottle 27227-2.5L-M which is physically identical with the same exact specifications.
The article number 27227-4X2.5L-R will be discontinued. Please order the single bottle 27227-2.5L-R which is physically identical with the same exact specifications.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Complete oxidation of CO, ethanol, and ethyl acetate over copper oxide supported on titania and ceria modified titania.
Larsson PO and Andersson A.
J. Catal., 179(1), 72-89 (1998)
K H Young et al.
Journal of clinical microbiology, 10(6), 852-853 (1979-12-01)
Ethyl acetate appears to be a satisfactory subsitute solvent for diethyl ether in the Formalin-ether sedimentation technique. In comparative studies, concentration of organisms with ethyl acetate was equal to or greater than that with diethyl ether. No distortion or alteration
K J S Anand et al.
British journal of anaesthesia, 101(5), 680-689 (2008-08-30)
Relationships between plasma morphine concentrations and neonatal responses to endotracheal tube (ETT) suctioning are unknown in preterm neonates. Ventilated preterm neonates (n=898) from 16 centres were randomly assigned to placebo (n=449) or morphine (n=449). After an i.v. loading dose (100
Yuusuke Fujimoto et al.
PloS one, 7(11), e48685-e48685 (2012-11-13)
Combination therapy with ribavirin, interferon, and viral protease inhibitors could be expected to elicit a high level of sustained virologic response in patients infected with hepatitis C virus (HCV). However, several severe side effects of this combination therapy have been
Maarit Neuvonen et al.
PloS one, 9(8), e103743-e103743 (2014-08-27)
Bacterial cholesterol oxidase is commonly used as an experimental tool to reduce cellular cholesterol content. That the treatment also generates the poorly degradable metabolite 4-cholesten-3-one (cholestenone) has received less attention. Here, we investigated the membrane partitioning of cholestenone using simulations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service