107603
Sodium formate
reagent grade, 97%
Synonym(s):
Formic acid sodium salt
About This Item
powder, crystals or granules
Recommended Products
grade
reagent grade
Assay
97%
form
crystalline powder
powder, crystals or granules
mp
259-262 °C (lit.)
solubility
water: soluble(lit.)
SMILES string
[Na+].[O-]C=O
InChI
1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1
InChI key
HLBBKKJFGFRGMU-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Engineering Redox Cofactor Balance for Improved 5-Methyltetrahydrofolate Production in Escherichia coli.: Sodium formate was used to manipulate redox cofactor balance, significantly enhancing the production of 5-methyltetrahydrofolate in E. coli, showcasing its potential in metabolic engineering applications (Yang et al., 2024).
- Intensity-dependent mass search for improving metabolite database matches in chemical isotope labeling LC-QTOF-MS-based metabolomics.: Sodium formate was utilized in isotope labeling for LC-QTOF-MS, improving metabolite database matching accuracy, thus enhancing metabolomics data quality (Li et al., 2023).
- High-Yield Synthesis of Enantiopure 1,2-Amino Alcohols from l-Phenylalanine via Linear and Divergent Enzymatic Cascades.: The study highlights sodium formate′s role in enzymatic cascades for the efficient synthesis of enantiopure amino alcohols, important intermediates in pharmaceutical synthesis (Corrado et al., 2022).
- UPLC-ESI-QTOF/MS and multivariate data analysis for blood plasma and serum metabolomics: effect of experimental artefacts and anticoagulant.: Sodium formate was used as a reference standard in UPLC-ESI-QTOF/MS, providing insights into minimizing experimental artefacts in blood plasma and serum metabolomics (Barri and Dragsted, 2013).
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service