Skip to Content
Merck
All Photos(1)

Documents

Y0000535

Terbinafine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

trans-N-(6,6-Dimethyl-2-hepten-4-ylyl)-N-methyl-1-naphthylmethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25N · HCl
CAS Number:
Molecular Weight:
327.89
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

terbinafine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].[H]\C(CN(C)Cc1cccc2ccccc12)=C(\[H])C#CC(C)(C)C

InChI

1S/C21H25N.ClH/c1-21(2,3)15-8-5-9-16-22(4)17-19-13-10-12-18-11-6-7-14-20(18)19;/h5-7,9-14H,16-17H2,1-4H3;1H/b9-5+;

InChI key

BWMISRWJRUSYEX-SZKNIZGXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Allylamine derivative.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Terbinafine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Mode of Action: Inhibits squalene epoxidase, preventing biosynthesis of ergosterol.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Jorge Casado et al.
Analytical and bioanalytical chemistry, 407(3), 907-917 (2014-09-28)
An effective and selective, modular sample preparation method for the extraction of eight antimycotic drugs, belonging to three different chemical classes, from digested sludge samples is proposed. To this end, matrix solid-phase dispersion (MSPD) was on-line connected with a cationic
Sylwia Magiera et al.
Journal of AOAC International, 97(6), 1526-1537 (2015-01-30)
A rapid and sensitive ultra-HPLC/MSIMS (UHPLC/MSIMS) assay method for the simultaneous determination in human urine of 22 drugs belonging to different pharmaceutical groups was developed. The drugs were extracted from urine samples and then separated on a Zorbax Rapid Resolution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service