Skip to Content
Merck
All Photos(1)

Key Documents

SMB00980

Sigma-Aldrich

Cortisol sulfate sodium salt

≥95% (HPLC)

Synonym(s):

Cortisol, 21-(hydrogen sulfate), monosodium salt (6CI, 7CI, 8CI), Sodium 11,17-dihydroxy-3,20-dioxopregn-4-en-21-yl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30NaO8S
CAS Number:
Molecular Weight:
465.51
UNSPSC Code:
51420000
NACRES:
NA.25
Assay:
≥95% (HPLC)
form:
solid

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

2-8°C

SMILES string

O=C(COS(O)(O[Na])=O)[C@@]1(O)CC[C@@]2([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@@]3([H])[C@@H](O)C[C@@]21C)=O

InChI

1S/C21H30O8S.Na/c1-19-7-5-13(22)9-12(19)3-4-14-15-6-8-21(25,17(24)11-29-30(26,27)28)20(15,2)10-16(23)18(14)19;/h9,14-16,18,23,25H,3-8,10-11H2,1-2H3,(H,26,27,28);/q;+1/p-1/t14-,15-,16-,18+,19-,20-,21-;/m0./s1

InChI key

DJKQHUMDQFOLHE-WDCKKOMHSA-M

General description

Cortisol sulfate is a urinary metabolite of steroid hormone cortisol. Cortisol sulfate was proposed as lung cancer biomarker. It is also reported at high levels in breast cancer, liver cancer and other cancers.

Application

Cortisol sulfate sodium salt, a derivative of the hormone cortisol, plays a crucial role in numerous physiological processes. Its applications in metabolomics extend to studying stress response, adrenal function, metabolism, and its links to conditions like obesity, diabetes, and psychiatric disorders. This versatile molecule is extensively utilized in biochemical and metabolomics research for its diverse applications.

Features and Benefits

  • High-quality molecule suitable for mulitple research applications
  • Commonly employed in Metabolomics and Biochemical studies

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Majda Haznadar et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 28(10), 1704-1711 (2019-07-31)
Liver cancer is the second leading cause of cancer-related deaths worldwide. With a predicted 2.4-fold rise in liver cancer incidence by 2020, there is an urgent need for early, inexpensive diagnostic biomarkers to deploy in the clinic. We employed ultraperformance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service