NIST2971
24R,25-Dihydroxyvitamin D3 Calibration Solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
packaging
ampule of 5 × 1 mL
manufacturer/tradename
NIST®
application(s)
vitamins, nutraceuticals, and natural products
format
single component solution
General description
This Standard Reference Material (SRM) is intended for use in the calibration of instruments and techniques used for the determination of 24R,25-dihydroxyvitamin D3. A unit of this product consists of five 2 mL ampoules of ethanolic solution.
Each ampoule contains approximately 1 mL of solution.
Each ampoule contains approximately 1 mL of solution.
Other Notes
Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
24R,25-Dihydroxyvitamin D3 [24R,25(OH)2D3]
See certificate for values and more details at nist.gov/SRM.
24R,25-Dihydroxyvitamin D3 [24R,25(OH)2D3]
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
55.4 °F
Flash Point(C)
13 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service