NIST2895
Ethanol-water solution
NIST® SRM® 2895, nominal mass fraction 0.2%
Synonym(s):
Ethanol solution, Ethyl alcohol
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 5 x 1.2 mL
manufacturer/tradename
NIST®
technique(s)
Karl-Fischer titration: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
single component solution
SMILES string
CCO
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3
InChI key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
SRM 2895_cert
SRM 2895_SDS
Other Notes
Ethanol (C2H6O)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service