Skip to Content
Merck
All Photos(1)

Key Documents

E8631

Supelco

3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulfonic acid sodium salt

for enzymic, spectrophotometric det. of H2O2

Synonym(s):

3-(N-Ethyl-m-toluidino)-2-hydroxypropanesulfonic acid sodium salt, N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-m-toluidine sodium salt, Sodium 3-(N-ethyl-3-methylanilino)-2-hydroxypropanesulfonate, Sodium N-ethyl-N-(2-hydroxy-3-sulfopropyl)-m-toluidine, TOOS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18NNaO4S
CAS Number:
Molecular Weight:
295.33
Beilstein:
5689521
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

quality

for enzymic, spectrophotometric det. of H2O2

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

[Na+].CCN(CC(O)CS([O-])(=O)=O)c1cccc(C)c1

InChI

1S/C12H19NO4S.Na/c1-3-13(8-12(14)9-18(15,16)17)11-6-4-5-10(2)7-11;/h4-7,12,14H,3,8-9H2,1-2H3,(H,15,16,17);/q;+1/p-1

InChI key

IRQRBVOQGUPTLG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamaoku, K., et al.
Chemical & Pharmaceutical Bulletin, 30, 2492-2492 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service