Skip to Content
Merck
All Photos(1)

Documents

B1143000

Bromazepam

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

7-Bromo-5-(2-pyridyl)-3H-1,4-benzodiaxepin-2(1H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10BrN3O
CAS Number:
Molecular Weight:
316.15
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

bromazepam

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=C(CN=C1C2=CC=CC=N2)NC3=C1C=C(Br)C=C3

InChI

1S/C14H10BrN3O/c15-9-4-5-11-10(7-9)14(17-8-13(19)18-11)12-3-1-2-6-16-12/h1-7H,8H2,(H,18,19)

InChI key

VMIYHDSEFNYJSL-UHFFFAOYSA-N

Gene Information

human ... GABRA2(2555)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bromazepam EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marlo Cunha et al.
Arquivos de neuro-psiquiatria, 67(2A), 214-218 (2009-06-24)
This study aimed to elucidate cortical mechanisms and to identify the areas where occur such mechanisms due to interaction between bromazepam and motor learning. The sample was composed of 45 healthy subjects randomly distributed in 3 groups: placebo (n=15), bromazepam
S Djezzar et al.
International journal of legal medicine, 123(3), 213-219 (2008-10-18)
Psychoactive substances may be administered without the knowledge of a victim in order to induce incapacitation and thus facilitate criminal actions. The characteristics of the victims and the drugs used in such suspected chemical submissions (CS) were analyzed in 309
M S Attia
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 74(4), 972-976 (2009-09-29)
A rapid, simple and sensitive spectrofluorimetric method for determination of trace amount of bromazepam is developed. In phosphate buffer of pH 7.4. The bromazepam enhance the luminescence intensity of the Eu(3+) ion in Eu(3+)-bromazepam complex at lambda(ex)=390nm. The produced luminescence
Marlo Cunha et al.
Neuroscience letters, 448(1), 33-36 (2008-10-22)
The aim of this study was to investigate the influence of bromazepam on EEG and the motor learning process when healthy subjects were submitted to a typewriting task. We investigated bromazepam due to its abuse by various populations and its
Yvan Gaillard et al.
Forensic science international, 207(1-3), e16-e18 (2011-01-05)
We reported on the death by poisoning of a one-month-old baby that had followed the death of one of her sister (due to cyamemazine overdose). Exhumation of the corpse was done 8 months after burial and revealed the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service