Skip to Content
Merck
All Photos(5)

Key Documents

906832

Sigma-Aldrich

Dimethyl isosorbide

greener alternative

BioRenewable, ReagentPlus®, ≥99%

Synonym(s):

Isosorbide dimethyl ether, DMI, 1,4:3,6-Dianhydro-2,5-di-O-methyl-D-glucitol, 1,4:3,6-Dianhydrosorbitol 2,5-dimethyl ether, 2,5-Di-O-methyl-1,4:3,6-dianhydro-D-glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O4
CAS Number:
Molecular Weight:
174.19
Beilstein:
80854
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.07
Assay:
≥99%
bp:
93-95 °C/0.1 mmHg (lit.)
Pricing and availability is not currently available.

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

liquid

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.461 (lit.)

bp

93-95 °C/0.1 mmHg (lit.)

density

1.15 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

CO[C@H]1COC2[C@@H](COC12)OC

InChI

1S/C8H14O4/c1-9-5-3-11-8-6(10-2)4-12-7(5)8/h5-8H,3-4H2,1-2H3/t5-,6+,7-,8-/m1/s1

InChI key

MEJYDZQQVZJMPP-ULAWRXDQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Dimethyl isosorbide (DMI) is a biobased greener solvent and thus it aligns with "Use of Renewable Feedstocks" and "Safer Solvents and Auxiliaries". Click here for more information. Dimethyl isosorbide (DMI) is a sustainable alternative to dipolar aprotic solvents such as NMP and DMF, suitable for organic synthesis, as Baylis-Hillman reaction, and due to its high solubilization capacity when used as co-solvent for nonpolar drugs, make it an interesting compound for pharmaceutical and cosmetic industries as well.

Application

Dimethyl isosorbide may be used as an alternative solvent to NMP and/or DMF in some applications:
  • Polymer membranes preparation
  • Solvent in Baylis-Hillman reactions
  • Co-solvent to improve solubility of nonpolar compounds
  • Potential substitutes to short-chain glycol ethers

Features and Benefits

  • Bio-based
  • Made from Renewable Resource - straw
  • Non-toxic solvent
  • High solubility potential
  • High boiling point

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Green Synthesis of Dimethyl Isosorbide
Pietro Tundo Prof. et al
ChemSusChem, 3, 566-570 (2010)

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Questions

1–5 of 5 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Why should you use BioRenewable solvents?

    1 answer
    1. You will contribute to create a sustainable world, significantly lowering environmental impact. Biorenewable solvents also decrease petroleum sources dependency, have higher recovery rates, and/or are safer for handling and disposal.

      Helpful?

  4. Do BioRenewable solvents have a similar quality to petroleum-based solvents?

    1 answer
    1. BioRenewable solvents can have the same or higher purity level compared to traditional solvents. These solvents are free of the many byproducts of petroleum manufacturing, such as benzene, aldehydes, and ethers.

      Helpful?

  5. What is a BioRenewable solvent?

    1 answer
    1. BioRenewable solvents are sourced from renewable and sustainable biobased materials, such as soy/palm oil, wood waste, hay or corn byproducts, etc. It is important to highlight that our BioRenewable products do not compete with food production.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service