Skip to Content
Merck
All Photos(1)

Key Documents

793213

Sigma-Aldrich

Reagent Alcohol

70%

Synonym(s):

Ethanol, Ethyl alcohol, EtOH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
15101802
NACRES:
NA.21

vapor density

1.59 (vs air)

Quality Level

form

liquid

expl. lim.

3.1-27.7 % (lit.)

concentration

70%

bp

78 °C (lit.)

mp

-114 °C (lit.)

Tax and license information

Sales restrictions may apply

Looking for similar products? Visit Product Comparison Guide

Application

Reagent alcohol has been used as a solvent in:
  • The SELEX (Systematic Evolution of Ligands by Exponential Enrichment) procedure for the selection of aptamers.
  • The study of the morphology of mitochondria in dopaminergic neurites of mice.

Packaging

4x1GA-PB is in poly bottle.
12x1GA-PB is 3x(4x1GA) in poly bottle.
20x1GA-PB is 5x(4x1GA) in poly bottle.

Components

95 parts of specially denatured ethyl alcohol 3A, 140 proof, with 5 parts of isopropyl alcohol. Final concentrations are ~ 60% ethanol, ~ 5% methanol and ~ 5% isopropanol. Contains ~ 30% water.

Other Notes

Ordering Information:
The letterhead approval is not required by Sigma-Aldrich for the 4x1GA unit size.

Link to Letterhead Example:

replacement part

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Image-Based Analysis of Mitochondrial Area and Counting from Adult Mouse Dopaminergic Neurites
Nissanka N, et al.
Bio-protocol, 8(16) (2018)
In vitro selection and amplification protocols for isolation of aptameric sensors for small molecules
Yang K-A, et al.
Methods, 106, 58-65 (2016)
Sonia Soloneski et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(5), 834-844 (2015-03-31)
In vitro effects of the carbamates pirimicarb and zineb and their formulations Aficida® (50% pirimicarb) and Azzurro® (70% zineb), respectively, were evaluated in Chinese hamster ovary (CHO-K1) cells. Whereas the cytokinesis-blocked micronucleus cytome assay was employed to test for genotoxicity
U Allenstein et al.
Soft matter, 11(25), 5053-5059 (2015-06-02)
Cells require adhesion to survive, proliferate and migrate, as well as for wound healing and many other functions. The strength of contractile cell forces on an underlying surface is a highly relevant quantity to measure the affinity of cells to
Nicholas J Niemuth et al.
Chemosphere, 135, 38-45 (2015-04-22)
The occurrence of intersex fish, where male reproductive tissues show evidence of feminization, have been found in freshwater systems around the world, indicating the potential for significant endocrine disruption across species in the ecosystem. Estrogens from birth control medications in

Articles

Cell counting protocol using a hemocytometer for monitoring cell viability. Discover other cell counting tools such as the Scepter™ 3.0 Cell Counter.

Cell counting protocol using a hemocytometer for monitoring cell viability. Discover other cell counting tools such as the Scepter™ 3.0 Cell Counter.

Cell counting protocol using a hemocytometer for monitoring cell viability. Discover other cell counting tools such as the Scepter™ 3.0 Cell Counter.

Cell counting protocol using a hemocytometer for monitoring cell viability. Discover other cell counting tools such as the Scepter™ 3.0 Cell Counter.

Protocols

Cell culture protocol for proper aseptic technique to maintain a sterile environment and to prevent cell culture contamination. Free ECACC handbook download.

Cell culture protocol for passaging and splitting adherent cell lines using trypsin EDTA. Free ECACC handbook download.

Cell culture protocol for passaging and splitting adherent cell lines using trypsin EDTA. Free ECACC handbook download.

Cell culture protocol for passaging and splitting adherent cell lines using trypsin EDTA. Free ECACC handbook download.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service