Recommended Products
product line
Selectophore™
form
liquid
composition
Methyltriphenylphosphonium bromide, 0.25 wt. % (130079)
Nitrocellulose, moistened with 2-propanol (35%), 1.93 wt. %
2-Nitrophenyl octyl ether, 16.25 wt. % (73732)
Poly(vinyl chloride), 5.75 wt. % (81392)
Tetrahydrofuran, 74.32 wt. % (87369)
Tridodecylmethylammonium nitrate, 1.50 wt. % (91664)
storage temp.
−20°C
General description
Application
Packaging
Other Notes
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
1.4 °F - closed cup
Flash Point(C)
-17.0 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service