Skip to Content
Merck
All Photos(2)

Key Documents

34010

Supelco

2-NP-AHD-13C3

VETRANAL®, analytical standard

Synonym(s):

1-(2-Nitrobenzylidenamino)-2,4-imidazolidinedione-[2,4,5-13C]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C7H8N4O4
CAS Number:
Molecular Weight:
251.17
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

mass shift

M+3

SMILES string

[O-][N+](=O)c1ccccc1\C=N\N2[13CH2][13C](=O)N[13C]2=O

InChI

1S/C10H8N4O4/c15-9-6-13(10(16)12-9)11-5-7-3-1-2-4-8(7)14(17)18/h1-5H,6H2,(H,12,15,16)/b11-5+/i6+1,9+1,10+1

InChI key

FULJCJZKZXOFQZ-OTQLJULOSA-N

Application

2-NP-AHD-13C3 may be used as a standard in the determination of nitrofuran metabolites and chloramphenicol in shrimp, using single extraction method and liquid chromatographic/mass spectrometric method (LC/MS-MS).
2-NP-AHD-13C3 may be used as an internal standard to quantify the presence of 1-(2-nitrobenzylidenamino)-2,4-imidazolidine-dione (2-NP-AHD) in shrimp samples using liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aref El-Demerdash et al.
Journal of AOAC International, 98(3), 595-601 (2015-05-31)
A method has been developed to quantify the nitrofuran metabolites 3-amino-5-morphorinomethyl-1,3-oxazolidinone, 3-amino-oxazolidinone, 1-aminohydantoin, and semicarbazide, as well as chloramphenicol in shrimp with a single extraction procedure followed by LC-MS/MS analysis. Dynamic selected reaction monitoring with positive and negative ionization mode
Simultaneous determination of nitrofuran metabolites and chloramphenicol in shrimp with a single extraction and LC-MS/MS analysis.
El-Demerdash A, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 98(3), 595-601 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service