Skip to Content
Merck
All Photos(1)

Documents

33383

Supelco

Triasulfuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16ClN5O5S
CAS Number:
Molecular Weight:
401.83
Beilstein:
7151883
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1nc(C)nc(NC(=O)NS(=O)(=O)c2ccccc2OCCCl)n1

InChI

1S/C14H16ClN5O5S/c1-9-16-12(19-14(17-9)24-2)18-13(21)20-26(22,23)11-6-4-3-5-10(11)25-8-7-15/h3-6H,7-8H2,1-2H3,(H2,16,17,18,19,20,21)

InChI key

XOPFESVZMSQIKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Triasulfuron is a pre-and post-emergence sulfonyl urea herbicide used for the selective weed control in barley and wheat.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Triasulfuron may be used as an analytical reference standard for the determination of the analyte in water and soil samples using micellar electro-kinetic capillary chromatography.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Pose-Juan et al.
Frontiers in microbiology, 8, 378-378 (2017-03-25)
The effect of organic amendments and pesticides on a soil microbial community has garnered considerable interest due to the involvement of microorganisms in numerous soil conservation and maintenance reactions. The aim of this work was to assess the influence on
Herbicides Inhibiting Branched-Chain Amino Acid Biosynthesis, 10 (2012)
Fabrice G Renaud et al.
Chemosphere, 57(10), 1525-1535 (2004-11-03)
Diffusion-retarded partitioning of pesticides with aggregated soils results in a time-dependent partition coefficient (Kd') which is different at equilibrium from the partition coefficient derived from conventional 24-h batch studies (Kd) measured on dispersed soil. An experiment was undertaken to determine
F A Macías et al.
Journal of agricultural and food chemistry, 48(7), 3003-3007 (2000-07-18)
A collection of 11 natural and synthetic podolactones have been tested as allelochemicals in a range between 10(-4) and 10(-9) M, and their potential use as natural herbicide models is discussed. Their effects on the germination and growth of the
Darryl R Nelson et al.
Antonie van Leeuwenhoek, 93(1-2), 123-132 (2007-07-13)
Most bacteria possess the enzyme acetohydroxyacid synthase, which is used to produce branched-chain amino acids. Enteric bacteria contain several isozymes suited to different conditions, but the distribution of acetohydroxyacid synthase in soil bacteria is largely unknown. Growth experiments confirmed that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service