Skip to Content
Merck
All Photos(1)

Key Documents

16447

Millipore

Glucose Bromcresol Purple Agar

NutriSelect® Plus, suitable for microbiology

Synonym(s):

Tryptone Yeast Extract Agar with Bromocresol Purple

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85
Pricing and availability is not currently available.

Agency

according to ISO 21258-1:2017

Quality Level

sterility

non-sterile

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

Agar, 15.0 g/L
Brom cresol purple, 0.015 g/L
Casein enzymic hydrolysate, 10.0 g/L
Glucose, 10.0 g/L
Sodium chloride, 5.0 g/L
Yeast extract, 1.5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | utilization test: suitable

final pH

7.0±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages

microbiology

suitability

Citrobacter spp.
Neisseria spp.
Proteus spp.
Serratia spp.
Shigella spp.
Streptococcus spp.
nonselective and differential for Bacillus cereus
nonselective and differential for Bacillus spp.
nonselective and differential for Enterobacter spp.
nonselective and differential for Escherichia coli
nonselective and differential for Klebsiella spp.
nonselective and differential for Salmonella spp.
nonselective and differential for coliforms
nonselective and differential for enterobacteriaceae

Application

Glucose Bromcresol Purple Agar is used for isolation and enumeration of Enterobacteriaceae and Bacillus cereus.

Preparation Note

Suspend 41.52 g in 1000 ml distilled water. Heat to boiling with gentle swirling to dissolve the agar completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Avoid overheating. Cool to 45-50°C and pour into sterile petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Microbiological assessment of bacteria found in spices, a potent source for food spoilage and pathogens.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

See All

Protocols

General guidance for the detection and enumeration of Enterobacteriaceae in food according to EN-ISO 8523:1991 and EN-ISO 4832:1991, respectively.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service