Skip to Content
Merck
All Photos(2)

Documents

05498

Supelco

Triphenyl phosphate

TraceCERT®, 31P-qNMR Standard, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)3PO
CAS Number:
Molecular Weight:
326.28
Beilstein:
1888236
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

vapor pressure

1.3 mmHg ( 200 °C)

Quality Level

product line

TraceCERT®

grade

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

qNMR: suitable

bp

244 °C/10 mmHg (lit.)

mp

48-50 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical

SMILES string

O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3

InChI

1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H

InChI key

XZZNDPSIHUTMOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Check out our entire range of quantitative NMR standards (qNMR standards)

Application


  • Triphenyl phosphate reagent in wastewater treatment: Triphenyl phosphate (TPP) is applied in advanced oxidation processes to enhance the removal of pollutants from wastewater, utilizing Myriophyllum biochar-supported Mn/Mg nano-composites as catalysts to activate periodate for efficient TPP degradation (Xie et al., 2024).

  • Organic phosphate compound in material science: TPP is utilized in the synthesis and modification of LDH hollow polyhedra by polydopamine, demonstrating its versatile application in materials science for developing new composite materials (Hou et al., 2024).

Other Notes

Ion Mobility: TWCCSN2 value of 169.2 Å2 [M+H]+ and 180.3 Å2 [M+Na]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 05498 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Recommended products

Check out ChemisTwin, our brand new online portal for identity confirmation of NMR spectra. A feature for quantitative NMR is in development. Learn more or reach out to us for a free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

428.0 °F - closed cup

Flash Point(C)

220 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Using high-performance quantitative NMR (HP-qNMR) for certifying traceable and highly accurate purity values of organic reference materials with uncertainties <0.1 %
Weber, M., Hellriegel, C., Rueck, A., Sauermoser, R., and Wuethrich, J.
Accreditation and Quality Assurance, 18, 91-98 (2013)
Michael Weber et al.
Analytical and bioanalytical chemistry, 407(11), 3115-3123 (2014-11-25)
Quantitative nuclear magnetic resonance (qNMR) spectroscopy is employed by an increasing number of analytical and industrial laboratories for the assignment of content and quantitative determination of impurities. Within the last few years, it was demonstrated that (1)H qNMR can be
Michael Weber et al.
Journal of pharmaceutical and biomedical analysis, 93, 102-110 (2013-11-05)
Quantitative NMR spectroscopy (qNMR) is gaining interest across both analytical and industrial research applications and has become an essential tool for the content assignment and quantitative determination of impurities. The key benefits of using qNMR as measurement method for the

Articles

The use of CRMs for the NMR quantification of phosphorylated organic compounds and metabolites offers several advantages as it is based on a signal comparison of the analyte with an internal or external reference standard.

Introduction of 19F qNMR certified reference materials for complex molecule certification.

Introduction of 19F qNMR certified reference materials for complex molecule certification.

Introduction of 19F qNMR certified reference materials for complex molecule certification.

See All

Related Content

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service