01378
Acetone-2,4-DNPH
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Acetone-2,4-dinitrophenylhydrazone
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
storage condition
under inert gas (argon)
technique(s)
gas chromatography (GC): suitable
mp
126-128 °C (lit.)
format
neat
storage temp.
2-8°C
SMILES string
C\C(C)=N\Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O
InChI
1S/C9H10N4O4/c1-6(2)10-11-8-4-3-7(12(14)15)5-9(8)13(16)17/h3-5,11H,1-2H3
InChI key
YGIXYAIGWMAGIB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Application
- High-throughput colorimetric assays for microbial production: Acetone-2,4-DNPH is utilized in high-throughput colorimetric assays, optimized for the detection of ketones and aldehydes in microbial cell factories, offering a robust method for monitoring biotechnological production processes (Kozaeva et al., 2022).
- Advanced detection of carbonyl compounds in airborne particles: Acetone-2,4-DNPH is employed in ultra-fast liquid chromatography coupled with mass spectrometry for the determination of free- and bound-carbonyl compounds in airborne particles, highlighting its critical role in environmental monitoring and health-related studies (Melo Cardozo et al., 2020).
- Quantitative diagnostic analysis of human blood: The compound is used in a method involving high-performance liquid chromatography for quantitative clinical diagnostic analysis of acetone in human blood, indicating its significance in metabolomic research and diagnostics (Akgul Kalkan et al., 2016).
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service