Skip to Content
Merck
All Photos(1)

Key Documents

28-4090

Sigma-Aldrich

Sodium selenite

SAJ first grade, ≥90.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SeO3
CAS Number:
Molecular Weight:
172.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

<0.00133 hPa ( 20 °C)

Assay

≥90.0%

form

powder

availability

available only in Japan

mp

>350 °C (lit.)

SMILES string

[Na+].[Na+].[O-][Se]([O-])=O

InChI

1S/2Na.H2O3Se/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

BVTBRVFYZUCAKH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Pacitti et al.
Aquatic toxicology (Amsterdam, Netherlands), 130-131, 97-111 (2013-02-07)
Selenium (Se) is an oligonutrient with both essential biological functions and recognized harmful effects. As the selenocysteine (SeCys) amino acid, selenium is integrated in several Se-containing proteins (selenoproteins), many of which are fundamental for cell homeostasis. Nevertheless, selenium may exert
Chih-Ching Yang et al.
PloS one, 9(7), e96006-e96006 (2014-07-02)
Deep-sea water (DSW), which is rich in micronutrients and minerals and with antioxidant and anti-inflammatory qualities, may be developed as marine drugs to provide intestinal protection against duodenal ulcers. We determined several characteristics in the modified DSW. We explored duodenal
Ying Yi Chen et al.
Cancer chemotherapy and pharmacology, 74(1), 25-35 (2014-05-08)
SN 28049 is a new DNA-binding topoisomerase II poison identified by its curative activity against the murine colon 38 carcinoma. Previous studies showed activity to be associated with selective drug accumulation and retention in tumour tissue. Retention varied widely among
Simon P W Hageman et al.
Water research, 47(7), 2118-2128 (2013-03-15)
Removal of the toxic selenium compounds selenite and selenate from waste water before discharge is becoming increasingly imperative in industrialized countries. Bacteria can reduce selenate to selenite, but also further to elemental selenium, selenide or organic selenium. In this paper
Abhishek Aggarwal et al.
Biochimica et biophysica acta, 1853(9), 2158-2167 (2015-02-24)
The inverse correlation between dietary calcium intake and the risk of colorectal cancer (CRC) is well known, but poorly understood. Expression of the calcium-sensing receptor (CaSR), a calcium-binding G protein-coupled receptor is downregulated in CRC leading us to hypothesize that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service