Skip to Content
Merck
All Photos(1)

Key Documents

24-1170

Sigma-Aldrich

Petrol

SAJ first grade, ≥90.0%

Synonym(s):

Gasoline, Petroleum benzin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
15101506
grade:
SAJ first grade
Assay:
≥90.0%

grade

SAJ first grade

Assay

≥90.0%

form

liquid

availability

available only in Japan

SMILES string

[Mg+]c1ccc(cc1)C.[Br-]

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gunnar Damgård Nielsen et al.
Basic & clinical pharmacology & toxicology, 98(2), 115-123 (2006-02-01)
Numerous studies have suggested that long-term occupational exposure to white spirit may cause chronic toxic encephalopathy (WHO 1996). This review summarizes the chronic nervous system effects of white spirit in animal studies during a 30-year period. First, routine histopathology was
S Edelfors et al.
Pharmacology & toxicology, 84(5), 197-200 (1999-06-11)
Female rats were exposed to white spirit (400 and 800 ppm for 6 hr/day) at day 7-20 during pregnancy. Thirty-five days after birth all female offspring were sacrificed, the brains removed, and the synaptosomal fractions prepared for in vitro studies.
Lena Ernstgård et al.
Journal of applied toxicology : JAT, 29(3), 255-262 (2008-12-17)
A move from 'standard' white spirit (stdWS, 15-20% aromatics) to low-aromatic or dearomatized white spirit (deWS) has been seen, as the latter are considered to carry a smaller risk of health effects. However, data on health risks of deWS on
H R Lam et al.
Toxicology letters, 80(1-3), 39-48 (1995-10-01)
The effects of 3 weeks' or 6 months' inhalation exposure of rats to aromatic white spirit 6 h/day, 5 days/week at 0, 400, or 800 ppm were studied. Synaptosomal neurochemistry was investigated as index of the in situ conditions in
S Dauger et al.
Archives de pediatrie : organe officiel de la Societe francaise de pediatrie, 10(3), 221-223 (2003-06-28)
The Authors report a case of acute White-Spirit poisoning with pulmonary hypertension associated to respiratory distress syndrome. A 14-month-old infant drank an unknown quantity of White-Spirit while his parents were painting. After he spontaneously vomited, he presented a seizure at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service