Skip to Content
Merck
All Photos(1)

Key Documents

15-2330

Sigma-Aldrich

2-Propanol

≥99.0%, SAJ first grade

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

product name

2-Propanol, SAJ first grade, ≥99.0%

grade

SAJ first grade

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

availability

available only in Japan

refractive index

n20/D 1.377 (lit.)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Selecting water-alcohol mixed solvent for synthesis of polydopamine nano-spheres using solubility parameter.: This research highlights the use of 2-propanol as a solvent in the synthesis of polydopamine nanoparticles, optimizing conditions for particle size and uniformity through solubility parameter techniques (Jiang et al., 2014).

  • Efficient enantioselective synthesis of (R)-[3,5-bis(trifluoromethyl)phenyl] ethanol by Leifsonia xyli CCTCC M 2010241 using isopropanol as co-substrate.: This study demonstrates the use of 2-propanol in the enantioselective synthesis of chiral alcohols, highlighting its role as a co-substrate in biocatalysis (Ouyang et al., 2013).

  • Real-time ESI-MS of enzymatic conversion: impact of organic solvents and multiplexing.: The research examines the impact of various organic solvents, including 2-propanol, on enzymatic reactions using real-time electrospray ionization mass spectrometry, offering insights into solvent effects on enzyme kinetics (Scheerle et al., 2012).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.1 °F

Flash Point(C)

11.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hadas Grossman et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(8), 3206-3216 (2015-04-30)
Granulosa cells support the developing oocytes and serve as transducers of the ovulatory stimulus induced by LH surge. Fyn kinase is expressed in granulosa cells, though its role in these cells has not been studied. In human embryonic kidney 293T
Jan Rybniker et al.
Nature communications, 6, 7659-7659 (2015-07-15)
Better antibiotics capable of killing multi-drug-resistant Mycobacterium tuberculosis are urgently needed. Despite extensive drug discovery efforts, only a few promising candidates are on the horizon and alternative screening protocols are required. Here, by testing a panel of FDA-approved drugs in
H Dolatshad et al.
Leukemia, 29(5), 1092-1103 (2014-11-28)
The splicing factor SF3B1 is the most commonly mutated gene in the myelodysplastic syndrome (MDS), particularly in patients with refractory anemia with ring sideroblasts (RARS). We investigated the functional effects of SF3B1 disruption in myeloid cell lines: SF3B1 knockdown resulted
Thomas W Winkler et al.
PLoS genetics, 11(10), e1005378-e1005378 (2015-10-02)
Genome-wide association studies (GWAS) have identified more than 100 genetic variants contributing to BMI, a measure of body size, or waist-to-hip ratio (adjusted for BMI, WHRadjBMI), a measure of body shape. Body size and shape change as people grow older
Swapna Ganapathy et al.
Journal of the American Chemical Society, 136(46), 16335-16344 (2014-10-24)
Fundamental research into the Li-O2 battery system has gone into high gear, gaining momentum because of its very high theoretical specific energy. Much progress has been made toward understanding the discharge mechanism, but the mechanism of the oxygen evolution reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service