Skip to Content
Merck
All Photos(1)

Key Documents

07-4872

Sigma-Aldrich

Dimethyl sulfoxide

≥99.0%, suitable for absorption spectrum analysis

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

availability

available only in Japan

technique(s)

UV/Vis spectroscopy: suitable

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

density

1.10 g/mL (lit.)

suitability

suitable for absorption spectrum analysis

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Oguro-Ando et al.
Molecular psychiatry, 20(9), 1069-1078 (2014-10-15)
Rare maternally inherited duplications at 15q11-13 are observed in ~1% of individuals with an autism spectrum disorder (ASD), making it among the most common causes of ASD. 15q11-13 comprises a complex region, and as this copy number variation encompasses many
Phillip McDonagh et al.
Veterinary microbiology, 174(3-4), 438-447 (2014-12-04)
Feline infectious peritonitis (FIP), a feline coronavirus (FCoV) induced disease, is almost invariably fatal with median life expectancy measured in days. Current treatment options are, at best, palliative. The objectives of this study were to evaluate a panel of nineteen
Claudia C Faria et al.
Cancer research, 75(1), 134-146 (2014-11-14)
Medulloblastoma is the most common malignant pediatric brain tumor, with metastases present at diagnosis conferring a poor prognosis. Mechanisms of dissemination are poorly understood and metastatic lesions are genetically divergent from the matched primary tumor. Effective and less toxic therapies
Rory W Blackler et al.
British journal of pharmacology, 172(4), 992-1004 (2014-10-10)
Hydrogen sulphide is an important mediator of gastrointestinal mucosal defence. The use of non-steroidal anti-inflammatory drugs (NSAIDs) is significantly limited by their toxicity in the gastrointestinal tract. Particularly concerning is the lack of effective preventative or curative treatments for NSAID-induced
Yan Xiang et al.
The Journal of clinical investigation, 125(6), 2293-2306 (2015-04-29)
Glutaminase (GLS), which converts glutamine to glutamate, plays a key role in cancer cell metabolism, growth, and proliferation. GLS is being explored as a cancer therapeutic target, but whether GLS inhibitors affect cancer cell-autonomous growth or the host microenvironment or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service