Skip to Content
Merck
All Photos(1)

Key Documents

03-5330

Sigma-Aldrich

γ-Butyrolactone

SAJ first grade, ≥99.5%

Synonym(s):

γ-Hydroxybutyric acid lactone, 4-Hydroxybutyric acid lactone, GBL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O2
CAS Number:
Molecular Weight:
86.09
Beilstein:
105248
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

SAJ first grade

vapor density

3 (vs air)

vapor pressure

1.5 mmHg ( 20 °C)

Assay

≥99.5%

autoignition temp.

851 °F

expl. lim.

16 %

availability

available only in Japan

refractive index

n20/D 1.436 (lit.)

bp

204-205 °C (lit.)

mp

−45 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCO1

InChI

1S/C4H6O2/c5-4-2-1-3-6-4/h1-3H2

InChI key

YEJRWHAVMIAJKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Precursor of γ-hydroxybutyric acid (GHB). It blocks dopamine release by blocking impulse flow in dopaminergic neurons. Pretreatment with γ-butyrolactone allows detection of autoreceptor-induced dopamine release.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michelle Wood et al.
Journal of chromatography. A, 1056(1-2), 83-90 (2004-12-15)
We have developed a rapid method that enables the simultaneous analysis of gamma-hydroxybutyrate (GHB) and its precursors, i.e. gamma-butyrolactone (GBL) and 1,4-butanediol (1,4-BD) in urine. The method comprised a simple dilution of the urine sample, followed by liquid chromatography-tandem mass
Christian Westendorf et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(10), 3853-3858 (2013-02-23)
The rapid reorganization of the actin cytoskeleton in response to external stimuli is an essential property of many motile eukaryotic cells. Here, we report evidence that the actin machinery of chemotactic Dictyostelium cells operates close to an oscillatory instability. When
Zheng-Yuan Su et al.
Chemical research in toxicology, 26(3), 477-485 (2013-02-28)
Cancer development has been linked to epigenetic modifications of cancer oncogenes and tumor suppressor genes; in advanced metastatic cancers, severe epigenetic modifications are present. We previously demonstrated that the progression of prostate tumors in TRAMP mice is associated with methylation
Bo Xue et al.
Biochemistry, 52(13), 2359-2370 (2013-03-07)
The in vitro evolution and engineering of quorum-quenching lactonases with enhanced reactivities was achieved using a thermostable GKL enzyme as a template, yielding the E101G/R230C GKL mutant with increased catalytic activity and a broadened substrate range [Chow, J. Y., Xue
Axel Bidon-Chanal et al.
European journal of medicinal chemistry, 60, 479-489 (2013-01-29)
Glycogen synthase kinase 3β (GSK-3β) is widely recognised as a relevant player in the pathogenesis of several highly prevalent disorders such as Alzheimer's disease, mood disorders, diabetes and cancer. Therefore, this enzyme constitutes a highly attractive therapeutic target for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service