Skip to Content
Merck
All Photos(1)

Documents

01-2690

Sigma-Aldrich

Aluminum chloride

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlCl3
CAS Number:
Molecular Weight:
133.34
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

1 mmHg ( 100 °C)

Assay

≥98.0%

form

solid

reaction suitability

reagent type: catalyst
core: aluminum

availability

available only in Japan

pH

2.4 (20 °C, 100 g/L)

mp

190 °C (lit.)

SMILES string

Cl[Al](Cl)Cl

InChI

1S/Al.3ClH/h;3*1H/q+3;;;/p-3

InChI key

VSCWAEJMTAWNJL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. Aluminum nitride can be synthesized from the elemnts with AlCl3 added to help facilitate crystallization.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diagnosis and management of hyperhidrosis.
R A Benson et al.
BMJ (Clinical research ed.), 347, f6800-f6800 (2013-11-28)
Bagineni Prasad et al.
Chemical communications (Cambridge, England), 48(84), 10434-10436 (2012-09-20)
A conceptually new and straightforward introduction of sulfonyl groups at the C-7 position of an indole ring has been achieved via AlCl(3) mediated unexpected regioselective sulfonyl group migration for N-alkyl/aryl/heteroarylsulfonyl indoles affording potential inhibitors of Mycobacterium tuberculosis H37Rv chorismate mutase.
Dharmalingam Prakash et al.
Neuromolecular medicine, 15(1), 192-208 (2013-01-15)
Aluminum (Al) is an environmental neurotoxin that affects cerebral functions and causes health complications. However, the role of Al in arbitrating glia homeostasis and pathophysiology remains obscure. Astrocyte, microglia activation (reactive gliosis), and associated inflammatory events play a decisive role
Luxin Zhang et al.
Bioresource technology, 130, 110-116 (2013-01-12)
In order to define a new green catalytic pathway for the production of furfural, the catalyzed conversion of xylan into furfural in 1-butyl-3-methylimidazolium chloride was studied by using mineral acids and metal chlorides as catalysts under microwave irradiation. Amongst these
Mengyun Zhu et al.
Organic letters, 16(7), 1856-1859 (2014-03-19)
An unprecedented AlCl3-promoted formal [2 + 3]-cycloaddition of 1,1-cyclopropanes with readily available N-benzylic sulfonamides has been developed. Experimental evidence supports an unusual mechanism wherein the donor-acceptor cyclopropane serves as a source of 2-styrylmalonate rather than the "classical" 1,3-dipole. A broad

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service