Skip to Content
Merck
All Photos(1)

Key Documents

XX1007615

Millipore

Microscope Slides

Accessories for filter holders for sample preparation.

Synonym(s):

Glass microscope slides, Laboratory slides

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

material

glass fiber slide

Quality Level

H

7.6 cm (3 in.)

W

5.0 cm (2 in.)

compatibility

for use with Patch Test Kit

shipped in

ambient

General description

A microscope slide is designed for the collection of various samples like the entire specimen, body parts of a specimen, and histological sections for both reference and research purposes. It has a significant role in history, veterinary, forensic, and medical fields. Microscope slide acts as an accessory for filter holders for sample preparation.

Application

Microscope slides have been used for histological studies of necrotic tissue and epithelial debris.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thomas Schmid et al.
Polymers, 13(13) (2021-07-03)
Microscope slide collections represent extremely valuable depositories of research material in a natural history, forensic, veterinary, and medical context. Unfortunately, most mounting media of these slides deteriorate over time, with the reason for this not yet understood at all. In
Hadrian Schepler et al.
Theranostics, 12(1), 18-34 (2022-01-07)
The healing of chronic wounds is impaired by a lack of metabolic energy. In previous studies, we showed that physiological inorganic polyphosphate (polyP) is a generator of metabolic energy by forming ATP as a result of the enzymatic cleavage of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service