Skip to Content
Merck
All Photos(1)

Key Documents

XX0020

Supelco

o-Xylene

Reagent

Synonym(s):

1,2-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
1815558
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-422-2
Assay:
≥98% (GC)
bp:
143-145 °C (lit.)
vapor pressure:
<0.1 atm ( 21.1 °C)
16 mmHg ( 37.7 °C)
7 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3.7 (vs air)

Quality Level

vapor pressure

<0.1 atm ( 21.1 °C)
16 mmHg ( 37.7 °C)
7 mmHg ( 20 °C)

Assay

≥98% (GC)

form

liquid

autoignition temp.

867 °F

expl. lim.

7 %

color

APHA: ≤10

refractive index

n20/D 1.505 (lit.)

bp

143-145 °C (lit.)

mp

-26--23 °C (lit.)

transition temp

flash point 30 °C

solubility

0.18 g/L

density

0.879 g/mL at 20 °C (lit.)
0.88 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

Cc1ccccc1C

InChI

1S/C8H10/c1-7-5-3-4-6-8(7)2/h3-6H,1-2H3

InChI key

CTQNGGLPUBDAKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Environmental sampling of volatile organic compounds during the 2018 Camp Fire in Northern California.: This study analyzed volatile organic compounds, including o-Xylene, sampled during the 2018 Camp Fire in Northern California, highlighting the analytical methods used to assess environmental exposure and chemical composition in wildfire scenarios (Simms et al., 2021).

  • Urinary BTEX, MTBE and naphthalene as biomarkers to gain environmental exposure profiles of the general population.: Focuses on the use of urinary biomarkers, such as BTEX (including o-Xylene), for assessing environmental exposure in populations, demonstrating the integration of analytical chemistry techniques in epidemiological studies (Fustinoni et al., 2010).

  • Genotoxic effects of volatile organic compounds in a chemical factory as evaluated by the Tradescantia micronucleus assay and by chemical analysis.: Investigates the genotoxic effects of VOCs, including o-Xylene, in a chemical plant environment, using both biological assays and chemical analysis to evaluate risk and exposure (Kim et al., 2003).

  • Time-gated pulsed glow discharge: real-time chemical speciation at the elemental, structural, and molecular level for gas chromatography time-of-flight mass spectrometry.: This paper presents a novel analytical technique for real-time chemical speciation, useful in analyzing compounds like o-Xylene, providing insights into complex analytical methodologies in chemistry (Lewis et al., 2003).

Analysis Note

Assay (GC): 98.0% min
Color (APHA): 10 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

  1. Merhaba, Ürün kaç derece sıcaklıkta gelicek? Ftalik anhidrit üretiminde kullanmak istiyorum. Tankta en fazla kaç derece sıcaklıkta bekletebilirim?

    1 answer
    1. This item is shipped and stored at room temp. Please see the link below to review a reference that may be helpful in termining the maximum temperature for the intended application:
      https://www.nist.gov/system/files/documents/srd/jpcrd412012the023103.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service