SX0665
Sodium Nitrite
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Synonym(s):
Sodium Nitrite, Diazotizing salts
About This Item
Recommended Products
grade
ACS reagent
Quality Level
Agency
meets USP/NF
Assay
≥97.0%
form
solid
impurities
≤0.01% Insoluble matter
pH
9 (20 °C, 100 g/L in H2O)
mp
280 °C (decomposition)
solubility
820 g/L
anion traces
chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.01%
cation traces
Ca: ≤0.01%
Fe: ≤0.001%
K: ≤0.005%
heavy metals (as Pb): ≤0.001%
shipped in
ambient
storage temp.
room temp
SMILES string
[Na+].N(=O)[O-]
InChI
1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1
InChI key
LPXPTNMVRIOKMN-UHFFFAOYSA-M
Application
Analysis Note
Calcium (Ca): 0.01% max
Chloride (Cl): 0.005% max
Heavy metals (as Pb): 0.001% max
Insoluble matter: 0.01% max
Iron (Fe): 0.001% max
Potassium (K): 0.005% max
Sulfate (SO4): 0.01% max
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service