Skip to Content
Merck
All Photos(1)

Key Documents

S-014-M

Sigma-Aldrich

Enzyme Free Cell Dissociation Solution PBS Based (1X), liquid, 500ml

The Enzyme-Free Cell Dissociation Solution is a PBS based formulation of chelating agents & agents to stabilize their activity on cells. This solution is available in a 500ml format.

Synonym(s):

Cell Dissociation Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160801
NACRES:
NA.75

Quality Level

form

liquid

manufacturer/tradename

Specialty Media

input

sample type hematopoietic stem cell(s)
sample type neural stem cell(s)
sample type pancreatic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type mesenchymal stem cell(s)
sample type cardiac stem cell(s)
sample type epithelial cells
sample type: mouse embryonic stem cell(s)
sample type: human embryonic stem cell(s)

General description

Enzyme-Free Cell Dissociation Solution is a PBS based proprietary formulation of chelating agents and agents to stabilize their activity on cells. The enzymatic activity of this formulation is comparable to trypsin. It preserves the structural and functional integrity of cell surface proteins and does not have the cytotoxic effects sometimes associated with 0.5 mM EDTA. It can be used to dissociate primary cells, tissues, and tumors with an increased plating efficiency.


Enzyme-Free dissociation solution should be used with cells that are sub-confluent (60-80%). If cells are more than 80% confluent, we recommend trypsinizing the cells and replating at 30-40% confluency 24 hours before enzyme-free use (this will allow the cells to be 60-80% confluent for enzyme free dissociation).

Physical form

Product is supplied as a sterile

liquid.

Storage and Stability

Product can be stored at room temperature or at 4°C. Shelf life is 2 years

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service