Skip to Content
Merck
All Photos(1)

Key Documents

NE1013

Sigma-Aldrich

Anti-PGP9.5 (176-191) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Neuron Cytoplasmic Protein 9.5, Anti-Ubiquitin Thiolesterase L1, Anti-UCHL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

porcine, human, rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UCHL1(7345)

General description

Rabbit polyclonal antibody supplied as undiluted serum. Recognizes the ~26 kDa PGP9.5 protein.
Recognizes the ~26 kDa PGP9.5 protein in mouse dorsal root nerve fibers.
This Anti-PGP9.5 (176-191) Rabbit pAb is validated for use in Immunoblotting, Frozen Sections for the detection of PGP9.5 (176-191).

Immunogen

a synthetic peptide (ASSEDTLLKDAAKVCR) corresponding to amino acids 176-191 of PGP9

Application


Immunoblotting (1:1000)
Frozen Sections (1:2000, fluorescence)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Mouse dorsal root nerve fibers

Other Notes

Caballero, O.L., et al. 2002. Oncogene21, 3003.
Tezel, E., et al. 2000. Clin. Cancer Res.6, 4764.
PGP9.5 is also overexpressed in some cancers.



Immunofluorescence protocol

Care should be taken so that the incubation solutions do not evaporate. It is recommended that incubations be performed in a humidified chamber to prevent evaporation. Washing steps should be done in large volumes of phosphate buffered saline (PBS).

1. Warm slides to room temperature.

2. Re-hydrate with PBS for 10-15 min.

3. Remove PBS and incubate in blocking buffer (PBS containing 3% normal donkey serum, 1% bovine serum albumin, 0.3% Triton X-100 detergent, and 0.01% NaN3, pH 7.45) for 1 h at room temperature.

4. Dilute primary antibody in blocking buffer. Store diluted antibody at 4°C until use.

5. Remove blocking buffer and add diluted primary antibody.

6. Incubate with primary antibody at 4°C for 18-48 h.

7. Remove primary antibody and wash three times for 10 min with PBS.

8. Dilute secondary antibody in blocking buffer to appropriate dilution (follow manufacture′s recommendation). Store at 4°C until use.

9. Incubate slides in diluted secondary antibody solution for one hour at room temperature.

10. Remove secondary antibody and wash three times for 10 min with PBS.

11. Coverslip slides with appropriate mounting media.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Nakano et al.
Physiological research, 61(6), 643-647 (2012-10-27)
This study was designed to investigate histological changes in skin tissue accompanying immobilization-induced hypersensitivity. Changes in mechanical sensitivity, epidermal thickness, and peripheral nerve profiles in the upper dermis were examined in glabrous skin of rat hind paw after 1, 2
Antonella S Ríos et al.
Development (Cambridge, England), 149(16) (2022-07-30)
The perception of noxious environmental stimuli by nociceptive sensory neurons is an essential mechanism for the prevention of tissue damage. Etv4 is a transcriptional factor expressed in most nociceptors in dorsal root ganglia (DRG) during the embryonic development. However, its
Masato Ogura et al.
PloS one, 16(2), e0246630-e0246630 (2021-02-05)
Interleukin-31 (IL-31) is involved in excessive development of cutaneous sensory nerves in atopic dermatitis (AD), leading to severe pruritus. We previously reported that PQA-18, a prenylated quinolinecarboxylic acid (PQA) derivative, is an immunosuppressant with inhibition of p21-activated kinase 2 (PAK2)
Sandra Boesmueller et al.
Journal of clinical medicine, 8(12) (2019-12-11)
The type II superior labrum anterior to posterior (SLAP) repair is a viable option in young and demanding patients, although a prolonged period of pain after surgery is described in the literature. The reason for this fact remains unknown. Thus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service