Skip to Content
Merck
All Photos(1)

Key Documents

MABT64

Sigma-Aldrich

Anti-Nectin-4/PVRL4 Antibody, clone N4.61

clone N4.61.2, from mouse

Synonym(s):

poliovirus receptor-related 4, Ig superfamily receptor LNIR, nectin 4, poliovirus receptor-related protein 4, nectin-4, Nectin-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N4.61.2, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PVRL4(81607)

General description

Nectin-4 is also known as poliovirus receptor-related protein 4 (PRR4 or PVRL4). Nectins are known for being the receptors for the herpes simplex virus. Nectin-4 is an immunoglobulin-like transmembrane cell adhesion protein that also seems to play a role in neural tube formation, with N-cadherin. Nectin-4 is connected with the actin cytoskeleton through afadin inside the cell. Nectin-4 is predominantly found in human placenta although in mice, expression is wide-ranging. Nectin-4 seems to adhere primarily to cells expressing nectin-1 and vice versa.

Immunogen

Recombinant protein corresponding to human Nectin-4.

Application

Anti-Nectin-4/PVRL4 Antibody, clone N4.61 is an antibody against Nectin-4/PVRL4 for use in IH, ELISA & IP.
ELISA Anaysis: A previous lot was used by an independent laboratory in ELISA (Fabre-Lafay, S., et al. (2005). Journal of Biological Chemistry. 280(20):19543–19550).

Immunoprecipitation Anaysis: A previous lot was used by an independent laboratory in IP (Fabre-Lafay, S., et al. (2005). Journal of Biological Chemistry. 280(20):19543–19550).
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated by Immunohistochemistry in ductal carcinoma tissue.

Immunohistochemistry Analysis: A 1:400 dilution of this antibody detected Nectin-4/PVRL4 in ductal carcinoma tissue.

Target description

55 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ Supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Ductal carcinoma tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristin L M Boylan et al.
Oncotarget, 8(6), 9717-9738 (2016-12-31)
The cell adhesion molecule Nectin-4 is overexpressed in epithelial cancers, including ovarian cancer. The objective of this study was to determine the biological significance of Nectin-4 in the adhesion, aggregation, migration, and proliferation of ovarian cancer cells. Nectin-4 and its
Justin W Kaufman et al.
Journal of virology, 97(10), e0105123-e0105123 (2023-09-21)
For many years, measles virus (MeV) was assumed to first enter the host via the apical surface of airway epithelial cells and subsequently spread systemically. We and others reported that MeV has an overwhelming preference for entry at the basolateral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service