Skip to Content
Merck
All Photos(1)

Key Documents

MABN790

Sigma-Aldrich

Anti-Shank3 Antibody, clone 16E2.3

clone 16E2.3, from mouse

Synonym(s):

SH3 and multiple ankyrin repeat domains protein 3, Shank3, Proline-rich synapse-associated protein 2, ProSAP2, Shank3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

16E2.3, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SHANK3(85358)

General description

SH3 and multiple ankyrin repeat domains protein 3 (UniProt Q9BYB0; also known as Proline rich synapse associated protein 2, ProSAP2, Shank postsynaptic density protein, Shank3) is encoded by the SHANK3 (also known as DEL22q13.3, PROSAP2, PSAP2, SCZD15, SPANK-2) gene (Gene ID 85358) in human. Shank3/ProSAP2 belongs to the ProSAP/Shank family of scaffolding proteins, which are important dendritic spines postsynaptic density (PSD) complex components that play an important role in linking membrane glutamate receptors to intracellular signaling proteins and actin cytoskeleton. Shank proteins interact with adaptor proteins and assembles glutamate receptors at the postsynaptic membrane via multiple domains, including ankyrin repeat, proline-rich, SH3, PSD95/disc-large/zona occludens-1 (PDZ), and SAM domains. Shanks, together with the adaptor protein Homer and the endocytic vesicle-associated protein dynamin-3, are required for proper localization of the endocytic zone in close proximity to the PSD. Shank-3 induces formation of functional dendritic spines in aspiny cerebellar neurons, whereas Shank-3 knockdown results in reduced spine density and size in cultured hippocampal neurons. SHANK3 gene mutations are known causes of mental retardation and autism spectrum disorders.

Specificity

Expected to react with both isoform 1 (A) and 2 (B).

Immunogen

Epitope: Internal (C-terminal half)
GST-tagged recombinant protein corresponding to the C-terminal half of human Shank3.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-Shank3 Antibody, clone 16E2.3 is validated for use in Western Blotting for the detection of Shank3.

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Shank3 in 10 µg of mouse brain tissue lysate.

Target description

~185 kDa observed. Multiplet banding pattern as a result of isoforms and/or posttranslational modifications. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service