Skip to Content
Merck
All Photos(1)

Key Documents

MABF369

Sigma-Aldrich

Anti-CD11c Antibody (human), PE-Cy7, clone 3.9

clone 3.9, from mouse

Synonym(s):

Integrin alpha-X, CD11 antigen-like family member C, Leu M5, Leukocyte adhesion glycoprotein p150, 95 alpha chain, Leukocyte adhesion receptor p150, 95, CD antigen CD11c

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

PE-Cy7

antibody form

purified antibody

antibody product type

primary antibodies

clone

3.9, monoclonal

species reactivity

human

packaging

pkg of 5 μL (1 μg/test)

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAX(3687)

General description

The 3.9 antibody reacts with human CD11c, also known as integrin alpha X. This 150 kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha X (CD11c) assembles with Integrin beta-2 (CD18) into a receptor known as CR4 which can bind and induce signaling through ICAMs and VCAM-1 on endothelial cells and can also facilitate removal of iC3b bearing foreign cells.The 3.9 antibody is widely used as a marker for CD11c expression on dendritic cells (DC), often in parallel with markers for CD11b, for identification of developmental stages and mature subsets of this cell type. CD11c is prominently expressed on tissue macrophages, and is also detected on activated neutrophils, granulocytes, some types of activated T cells and intestinal intraepithelial lymphocytes (IEL). The antibody is reported to be cross-reactive with Baboon, Chimpanzee, Cynomolgus and Rhesus CD11c.

Application

This Mouse Monoclonal Antibody Anti-CD11c (human), PE-Cy7, clone 3.9, Cat. No. MABF369 is validated for use in IP, IF, FC for the detection of CD11c.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service